Learn More
BRAND™ Filter Crucibles
Borosilicate glass construction. Squat form offers stability and low risk of spillage.
Brand: BRAND™ 458734
431.03 EUR valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Additional Details : Weight : 4.97000kg
Description
- Borosilicate glass construction
- Porosity Level: 4
- Supplied in packs of 10
Specifications
40 mm | |
50 mL | |
4 |
Borosilicate Glass | |
Filter crucible |