Learn More
Agilent BioTek Filter Cube
GREENER_CHOICE
Product Code. 15998550 Shop All Agilent BioTek Products
Change view
Click to view available options
Excitation/Emission:
EX 360/40, EM 460/40, 400 nm mirror (Blue)
EX 485/20, EM 528/20, 510 nm mirror (Green)
EX 530/25, EM 590/25, 570 nm mirror (Red)
Unit Size:
1 unit
Product Code. Excitation/Emission unitSize
15998550 EX 360/40, EM 460/40, 400 nm mirror (Blue) 1 unit
15988550 EX 485/20, EM 528/20, 510 nm mirror (Green) 1 unit
15978550 EX 530/25, EM 590/25, 570 nm mirror (Red) 1 unit
3 options
Product Code. 15998550
Brand: Agilent BioTek 1505006
For use with Synergy LX
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Synergy LX
Excitation/Emission EX 360/40, EM 460/40, 400 nm mirror (Blue)