missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Filter Disc, Untreated, Ground Cylindrical, 60 mm

The faces of the VitraPOR™ standard filter discs are press moulded and may show thickness deviations.

Brand:  ROBU VitraPOR™ 15604

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12040442

  • € 60.88 / Pack of 5


Estimated Shipment: 24-08-2021
Add to basket

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

60 mm
4
Borosilicate Glass 3.3
5/Pk.
5.0 ± 1.1 mm
Non-sterile
Videos
SDS
Documents
Certificates