missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Filter Disc, Untreated, Ground Cylindrical, 90 mm

The faces of the VitraPOR™ standard filter discs are press moulded and may show thickness deviations.

Brand:  ROBU VitraPOR™ 15903

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12024837

  • € 112.00 / Pack of 5


Estimated Shipment: 19-08-2021
Add to basket

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

90 mm
3
Borosilicate Glass 3.3
5/Pk.
6.5 ± 1.5 mm
Non-sterile
Videos
SDS
Documents
Certificates