missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Filter Disc, Untreated, Ground Cylindrical, 90 mm

The faces of the VitraPOR™ standard filter discs are press moulded and may show thickness deviations.

Brand:  ROBU VitraPOR™ 15900

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12014717

  • € 148.00 / Pack of 5
Estimated Shipment: 28-05-2024
to see stock.
Add to basket

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

Borosilicate Glass (3.3)
5/Pk.
6.5 ± 1.5 mm
Non-sterile
0
90 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers