Learn More
UVP Filter excitation for manual BioLite 600nm to 645nm UVP
Brand:  UVP 38-0359-03
Additional Details : Weight : 0.20000kg
Product Code. 12873069
  • € 670.00 / Each
Estimated Shipment: 12-07-2024