Learn More
UVP Filter excitation for manual BioLite 687nm to 748nm UVP
Brand:  UVP 38-0360-03
Additional Details : Weight : 0.20000kg
Product Code. 12883069
  • € 670.00 / Each
Estimated Shipment: 12-07-2024