Learn More
Witeg™ Filtration Flask with Plastic Hose Connection and Tubulature
Product Code. 10041732
Product Code. 10041732
Brand: Witeg™ 5 071 900
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Collect filtrate in these highly durable flasks. Witeg™ Filtration Flask with Plastic Hose Connection and Tubulature is made of highly resistant Duran™ with a Levasint™ safety coating.
  • Hose connection and tubulature.
  • Duran™ construction.
  • Levasint™ safety coating.
Specifications
Capacity (Metric) Media 10,000 mL
Includes Plastic hose connection and tubulature