missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ fimA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144499
This item is not returnable.
View return policy
Description
fimA Polyclonal Antibody for Western Blot, ELISA
Fimbriae (also called pili), polar filaments radiating from the surface of the bacterium to a length of 0.5-1.5 micrometers and numbering 100-300 per cell, enable bacteria to colonize the epithelium of specific host organs.
Specifications
| fimA | |
| Polyclonal | |
| Unconjugated | |
| fimA | |
| pilA | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| 948838 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 2.93 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| P04128 | |
| fimA | |
| Recombinant Escherichia coli Type-1 fimbrial protein, A chain (fimA) (24-182AA). | |
| 100 μL | |
| Primary | |
| Bacteria | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction