Learn More
Epredia™ Fine Pore Cassettes in Tube Packs
Product Code. 16384076 Shop All Epredia Products
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 750
Product Code. Color unitSize
16384076 Lavender Pack of 750
16344076 Beige Pack of 750
16354076 Blue Pack of 750
16364076 Green Pack of 750
16374076 Gray Pack of 750
16394076 Light Green Pack of 750
16304086 Lilac Pack of 750
16314086 Orange Pack of 750
16324086 Pink Pack of 750
16334086 Tan Pack of 750
16344086 White Pack of 750
16354086 Yellow Pack of 750
12 options
Product Code. 16384076
Brand: Epredia™ B851734LA
For use with very small biopsy tissue samples. These cassettes are compatible with all Epredia PrintMate™ Cassette Printers, have a front-edge lid hinge, and attached lids that are available in 12 color varieties.
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Fine Pore
Closure Included Yes
Quantity 750/Cs.
Closure Type Attached Lid, Front Hinge
PrintMate Compatible Yes
Color Lavender
For Use With (Equipment) For Use With PrintMate™ Printers
Product Type Cassette