Learn More
Thermo Scientific™ Finger Tite Ferrules
Product Code. 10055914 Shop All Thermo Scientific Products
Click to view available options
Diameter (Metric):
0.4 mm
0.5 mm
Unit Size:
Pack of 10
Product Code. 10055914
Brand: Thermo Scientific™ 290S1131
Thermo Scientific™ Finger Tite Ferrules offer easier fit, reliabililty and leak-free connections.
Thermo Scientific™ Finger Tite Ferrules offer easier fit, reliabililty and leak-free connections.
Thermo Scientific Finger Tite ferrules are the next generation of the SilTite™ connection system.
Advantages over conventional graphite/polymer ferrules:
  • Perfect metal-to-metal seal eliminates contamination and gives lower air/water background
  • No need to retighten ferrules, as they expand and contract with the fitting over GC operating temperatures
  • No tools required
TRUSTED_SUSTAINABILITY
Specifications
Type Ferrule
Description 0.5 mm ferrule
Diameter (Metric) 0.5 mm
For Use With (Equipment) 0.32 mm I.D. GC Columns