missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FIP1L1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-74589AF532
This item is not returnable.
View return policy
Description
FIP1L1 Polyclonal antibody specifically detects FIP1L1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| FIP1L1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| DKFZp586K0717, EC 1.1.1.86, EC 4.2.1.33, Factor interacting with PAP, FIP1, FIP1 like 1 (S. cerevisiae), FIP1-like 1 protein, FLJ33619, hFip1, pre-mRNA 3'-end-processing factor FIP1, Rearranged in hypereosinophilia, Rhe | |
| The immunogen recognized by this antibody maps to a region between residue 544 and 594 of human factor interacting with PAP using the numbering given in entry NP_112179.2 (GeneID 81608). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 81608 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction