missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FIP200 Monoclonal Antibody (2C9A11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA548675
This item is not returnable.
View return policy
Description
FIP200 Monoclonal Antibody for Western Blot, ELISA
The protein encoded by this gene interacts with signaling pathways to coordinately regulate cell growth, cell proliferation, apoptosis, autophagy, and cell migration. This tumor suppressor also enhances retinoblastoma 1 gene expression in cancer cells. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
Specifications
| FIP200 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q8TDY2 | |
| Rb1cc1 | |
| Purified recombinant fragment of human RB1CC1 (AA: 148-244) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot, Immunohistochemistry, Western Blot | |
| 2C9A11 | |
| Unconjugated | |
| Rb1cc1 | |
| 200 kDa FAK family kinase-interacting protein; 2900055E04Rik; 5930404L04Rik; ATG17; Cc1; coiled coil forming protein 1; coiled-coil-forming protein 1; FAK family kinase-interacting protein of 200 kDa; FIP200; KIAA0203; LaXp180; LOW QUALITY PROTEIN: RB1-inducible coiled-coil protein 1; phosphatase 1, regulatory subunit 131; PPP1R131; RB1 inducible coiled-coil 1; Rb1cc1; RB1-inducible coiled-coil 1; RB1-inducible coiled-coil protein 1; RBICC | |
| Mouse | |
| Protein G | |
| RUO | |
| 9821 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction