Learn More
Invitrogen™ FIP200 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA569698
Product Code. 15898103
  • € 530.00 / 100µL
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 100%.
The protein encoded by this gene interacts with signaling pathways to coordinately regulate cell growth, cell proliferation, apoptosis, autophagy, and cell migration. This tumor suppressor also enhances retinoblastoma 1 gene expression in cancer cells. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FIP200
Polyclonal
Unconjugated
Rb1cc1
200 kDa FAK family kinase-interacting protein; 2900055E04Rik; 5930404L04Rik; ATG17; Cc1; coiled coil forming protein 1; coiled-coil-forming protein 1; FAK family kinase-interacting protein of 200 kDa; FIP200; KIAA0203; LaXp180; LOW QUALITY PROTEIN: RB1-inducible coiled-coil protein 1; phosphatase 1, regulatory subunit 131; PPP1R131; RB1 inducible coiled-coil 1; Rb1cc1; RB1-inducible coiled-coil 1; RB1-inducible coiled-coil protein 1; RBICC
Rabbit
Affinity chromatography
RUO
9821
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry, Western Blot, Immunocytochemistry
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q8TDY2
Rb1cc1
synthetic peptide directed towards the C terminal region of human FIP200.