Learn More
Invitrogen™ FIR Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104510
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total FIR.
FARP2 (FERM, RhoGEF and pleckstrin domain-containing protein 2), also known as PLEKHC3 or FERM domain including RhoGEF (FIR), is a 1,545 amino acid protein that contains one FERM domain, one DH domain and two PH domains. It exists as two alternatively spliced isoforms that are abundantly expressed in brain, lung, and testis as well as in embryonic hippocampal and cortical neurons. FARP2 functions as a Rho-guanine nucleotide exchange factor that activates RAC1 and is thought to regulate neurite remodeling of embryonic neurons. Sema3A binding to neuropilin-1 induces the dissociation of FARP2 from plexin-A1, thereby activating FARP2's Rac GEF activity which is critical for repulsion of outgrowing axons and suppression of neuronal adhesion. Downregulation of the FARP2 gene has been implicated in autism.
Specifications
| FIR | |
| Polyclonal | |
| Unconjugated | |
| FARP2 | |
| AI465173; BC009153; D030026M03Rik; Farp2; FERM domain including RhoGEF; FERM domain-including RhoGEF; FERM, ARH/RhoGEF and pleckstrin domain protein 2; FERM, ARHGEF and pleckstrin domain-containing protein 2; FERM, RhoGEF and pleckstrin domain protein 2; FERM, RhoGEF and pleckstrin domain-containing protein 2; FGD1-related Cdc42-GEF; Fir; FRG; Kiaa0793; mKIAA0793; PH domain-containing family C member 3; pleckstrin homology domain-containing family C member 3; PLEKHC3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 227377, 9855 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O94887, Q91VS8 | |
| FARP2 | |
| A synthesized peptide derived from human FARP2(Accession O94887), corresponding to amino acid residues S344-L394. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |