Learn More
FIS1 Mouse anti-Human, CoraLite 488, Clone: 2F10E1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66635-100UL
Description
The FIS1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FIS1. This antibody recognizes human antigen. The FIS1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
| FIS1 | |
| Monoclonal | |
| 2000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Immunofluorescence | |
| 2F10E1 | |
| CoraLite 488 | |
| 100 μL | |
| 51024 | |
| IgG1 |