Learn More
FIS1 Mouse anti-Human, CoraLite 488, Clone: 2F10E1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-66635-100UL
Product Code. 16840826
  • € 450.00 / 100µL
Description
The FIS1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FIS1. This antibody recognizes human antigen. The FIS1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
Specifications
FIS1
Monoclonal
2000 μg/mL
Mouse
Primary
Human
Immunofluorescence
2F10E1
CoraLite 488
100 μL
51024