missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FIS1 Recombinant Rabbit Monoclonal Antibody (JE40-90)
GREENER_CHOICE

Product Code. 17135493
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17135493 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17135493 Supplier Invitrogen™ Supplier No. MA534822

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive Control: SK-Br-3, MCF-7, SH-SY-5Y, human kidney tissue, human small intestine tissue, human uterus tissue.

Fis1, a 17KDa integral membrane protein, is a novel member of mitochondrial fission machinery. It consists of a central leucine-zipper domain, a coiled-coil region, a tetratricopeptide repeat domain and a C-terminal transmembrane domain. It is a human orthologue of Fis1p (S.cerevisiae) and is localized in the outer mitochondrial membrane. Fis1 acts as a pro-apoptotic factor and plays a vital role in the regulation of mitochondrial fission. It interacts with DLP1 and Mdv1p and facilitates Dnm1p recruitment to the functional fission complex, thereby inducing apoptosis. Role of Fis1 has also been implicated in growth and division of peroxisomes. Loss of Fis1 inhibits apoptosis and results in elongation of mammalian mitochondria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FIS1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE40-90
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene FIS1
Gene Accession No. Q9Y3D6
Gene Alias 2010003O14Rik; CGI-135; Fis1; fis1 {ECO:0000250; FIS1 homolog; fission 1 (mitochondrial outer membrane) homolog; fission 1 (mitochondrial outer membrane) homolog (S. cerevisiae); fission 1 (mitochondrial outer membrane) homolog (yeast); fission, mitochondrial 1; H_NH0132A01.6; hFis1; Mitochondrial fission 1 protein; mitochondrial fission molecule; rFis1; tetratricopeptide repeat domain 11; tetratricopeptide repeat protein 11; TPR repeat protein 11; TTC11; UniProtKB:Q9Y3D6}
Gene Symbols FIS1
Host Species Rabbit
Immunogen Recombinant protein within Human TTC11 aa 1-125.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51024
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.