missing translation for 'onlineSavingsMsg'
Learn More

Fisetin, MedChemExpress

Product Code. 16412366
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16412366 500 mg 500mg
16409026 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16412366 Supplier Medchem Express Supplier No. HYN0182
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Fisetin is a natural flavonol found in many fruits and vegetables with various benefits, such as antioxidant, anticancer, neuroprotection effects.

Fisetin is a natural flavonol found in many fruits and vegetables with various benefits, such as antioxidant, anticancer, neuroprotection effects.

Specifications

Chemical Name or Material Fisetin
CAS 528-48-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H10O6
Quantity 500 mg
Solubility Information DMSO : 25 mg/mL (87.34 mM; Need ultrasonic)
SMILES O=C1C(O)=C(C2=CC=C(O)C(O)=C2)OC3=CC(O)=CC=C13
Molecular Weight (g/mol) 286.24
Formula Weight 286.24
Percent Purity 96.74%
Grade Research
Physical Form Solid powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.