Learn More
Fisherbrand™ accumet™ pH Indicating Glass Body Universal Glass Membrane Models- Mercury-Free
Universal glass membrane and glass body provides measurements from zero to 14pH
€ 169.95 - € 239.00
Product Code | Brand | Connector Types | Electrode | Length (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Connector Types | Electrode | Length (English) | Price | Quantity & Availability | |||
11550104
|
Fisherbrand™
|
BNC | Suggested Reference Electrodes: 13-620-51, 13-620-52, 13-620-57, 13-620-61, 13-620-62, 13-620-45, 13-620-53, 13-620-46, 13-620-47 | 30 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11593965
|
Fisherbrand™
|
U.S. Standard | Suggested Reference Electrodes: 13-620-64 | 60 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Universal glass membrane and glass body provides measurements from zero to 14pH
- Universal glass membrane
- Zero to 14pH range
Specifications
Glass | |
0 to 14 |