Learn More
Fisherbrand™ 25-Tooth Comb for Owl™ P2-CST Multiple Gradient Gel Caster
Product Code. 11913065 Shop All Thermo Scientific Products
Click to view available options
Teeth Count:
25
Unit Size:
Each
Product Code. 11913065
Brand: Fisherbrand™ P225C
Only null left
Add to basket
Add to basket
25-Tooth Comb for Owl™ P2-CST Multiple Gradient Gel Caster
  • For use with Owl P2-CST Multiple Gradient Gel Caster
  • Teeth count: 25
Note: Image is representative only. Exact number of teeth in photo may vary.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl P2-CST Multiple Gradient Gel Caster
Type 25-Tooth Comb
Gel Dimensions 20 x 20 cm
Teeth Count 25
Thickness (Metric) Teeth 0.08 cm
Volume (Metric) Well 69 μL
Length (English) Footprint 9.84 in.
Length (English) Gel 7.8 in.
Length (Metric) Footprint 25 cm
Length (Metric) Gel 20 cm
Width (English) Gel 7.8 in.
Width (English) Teeth 0.15 in.
Width (Metric) Footprint 13 cm
Width (Metric) Gel 20 cm
Width (Metric) Teeth 0.39 cm
Height (English) Footprint 9.05 in.
Height (Metric) Footprint 23 cm