Learn More
Fisherbrand™ Combs for FB-VE16-1 Vertical Electrophoresis System
Product Code. 11913055
Click to view available options
Volume (Metric) Well:
51 μL
69 μL
96 μL
107 μL
129 μL
183 μL
201 μL
343 μL
Length (Metric) Footprint:
16 cm
20 cm
Teeth Count:
10
15
20
24
Width (Metric) Teeth:
0.29 cm
0.39 cm
0.61 cm
1.04 cm
Unit Size:
Each
Product Code. 11913055
Brand: Fisherbrand™ P124D
Only null left
Add to basket
Add to basket
Creates the wells in gels for electrophoresis
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl P1-CST Multiple Gradient Gel Caster
Type Comb
Gel Dimensions 16 x 14 cm
Teeth Count 24
Thickness (Metric) Teeth 0.15 cm
Volume (Metric) Well 96 μL
Length (English) Footprint 6.29 in.
Length (English) Gel 6.3 in.
Length (Metric) Footprint 16 cm
Length (Metric) Gel 16 cm
Width (English) Gel 5.5 in.
Width (English) Teeth 0.11 in.
Width (Metric) Footprint 13 cm
Width (Metric) Gel 14 cm
Width (Metric) Teeth 0.29 cm
Height (English) Footprint 7.08 in.
Height (Metric) Footprint 18 cm