Learn More
Bio Plas™ Astral™ Inoculation Device System
Shop All Thermo Scientific Products
Click to view available options
Product Type:
Inoculating Loop
Color:
Yellow
Unit Size:
Each
Description
Includes
Autoclavable handle with every case
Economical, disposable inoculation system designed for safety and dependable sterility
Calibrated using Evans Blue Dye spectrophotometric procedure and gravimetric method
- Free of DNA, RNase, DNase, pyrogens and PCR inhibitors
- Astral loops, needles, and Clone-Piks are packaged static free into individual tubular chambers, which are then hermetically sealed with aluminum film
- A dust cover is heat-shrunk onto the unit before it is irradiation sterilized
- Use on an individual basis, or for quantitative procedures such as sampling, urine counts, or serial dilutions
To use:
- Puncture aluminum film with the handle, pushing down until it attaches to the loop, needle, or Clone-Pik
- Withdraw handle; inoculation device is ready for many applications
Order Info
Supplied 10 racks of 96 to a case
Specifications
Specifications
| Disposable | Single-use |
| Volume | 1 μL |
| Sterility | Sterile |
| Color | Yellow |
| Product Type | Inoculating Loop |
| For Use With (Equipment) | Astral™ inoculation device system |
| Includes | Autoclavable handle |
| Packaging Type | Rack |