Learn More
Fisherbrand™ Bags with Round-Wire Closure
Product Code. 17933182 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Dimensions (L x W):
5 x 3 in.
7 x 3 in.
9 x 4.5 in.
9 x 5.5 in.
12 x 4.5 in.
Thickness (English):
2.50 mil
3 mil
3.50 mil
4 mil
Capacity (English):
2 oz.
4 oz.
18 oz.
24 oz.
27 oz.
Color:
Clear
Clear with White Block
Unit Size:
Pack of 500
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Thickness (English) Capacity (English) Color Unit Size
17933182 9 x 4.5 in. 3 mil 18 oz. Clear with White Block Pack of 500
11924385 7 x 3 in. 3.50 mil 4 oz. Clear Pack of 500
17913182 5 x 3 in. 2.50 mil 2 oz. Clear Pack of 500
11964405 12 x 4.5 in. 3 mil 27 oz. Clear Pack of 500
11904395 9 x 4.5 in. 3 mil 18 oz. Clear Pack of 500
11944395 9 x 5.5 in. 3.50 mil 24 oz. Clear Pack of 500
11692549 - - - - -
11954385 7 x 3 in. 4 mil 4 oz. Clear with White Block Pack of 500
11944405 5 x 3 in. 2.50 mil 2 oz. Clear with White Block Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 17933182
Save Up to 60% on Liquid Handling
Use promo code 28028
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Secure, contaminant-free, pliant container ensures dependable analysis results
  • Secure
  • Contaminant-free
  • Pliant
Specifications
Closure Type Round Wire
Color Clear with White Block
Material Polyethylene
Sterility Sterile
Thickness (English) 3 mil
Dimensions (L x W) 9 x 4.5 in.
Length (English) 9 in.
Width (English) 4.5 in.
Capacity (English) 18 oz.
Length (Metric) 22.9 cm
Width (Metric) 11.4 cm