Learn More
Fisherbrand™ Combs for SUB-GEL Mini Horizontal Gel Electrophoresis Units, 0.75mm Thick
For use with SUB-GEL Mini Horizontal Gel Electrophoresis Units.
Brand: Fisherbrand™
Additional Details : Weight : 0.01000kg
Description
- Recommended for high resolution gels and publication quality data
- For tightly resolved bands
- Height-adjustable without any requirement for specialist tools or comb holders, to give user full control over well depth and sample-loading volume
- Rigid comb back prevents heat-induced warping
- Reversible loading guides sit directly above each well to provide a convenient loading template for single- and multi-channel pipets
- 8-sample and 12-sample combs are Multichannel pipettor compatible
Specifications
Black | |
Comb | |
1 Each | |
36 μL |
Sub-Gel Mini Horizontal Gel Electrophoresis Unit | |
4 Prep, 2 Marker | |
0.75 mm |