missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Cover Glasses: Circles

Made of finest optical borosilicate glass, with uniform thickness and size

Brand:  Fisherbrand™

Additional Details : Weight : 0.99000kg

 View more versions of this product

Product Code. 10319303

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Made of finest optical borosilicate glass, with uniform thickness and size

Circular shape.

  • Corrosion-resistant
  • Lint-free and ready to use
  • Available in squares, circles and rectangles

Order Info

Packed in 1 oz. (28g) plastic boxes with custom-molded bases for easy one-at-a-time dispensing

Specifications

Specifications

Coverslip
Borosilicate Glass
12 mm
Circle
#1.5 (0.16 to 0.19 mm)
Microscopes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers