Learn More
Fisherbrand™ Micryo™ Strips and Dots for Cryo Storage, Laser Printer Sheets, 0.5 x 1.25 in.; 85 labels/sheet; White
Intended uses of cryostorage with liquid nitrogen, refrigeration and ultra-low temperature freezer applications.
Brand: Fisherbrand™ 15-930-C
Includes: Template for printing instructions.
Description
Intended uses of cryostorage with liquid nitrogen, refrigeration and ultra-low temperature freezer applications.
Ideal for all cryostorage, liquid nitrogen, refrigeration, and ultra-low temperature freezer applications. Suitable for use on microcentrifuge tubes, cryovials, racks, boxes and microtiter plates. Adhesive withstands temperatures from -196° to 121°C (-321° to 250°F)
- No need to self-laminate
- Thinner labels allow tubes to fit in most storage systems
- Available in a full range of colors and sizes
- Guaranteed not to fall off under normal use (provided user instructions are followed)
- Also offered in rolls (see Cat. Nos. 15-910 and 15-920 Series)
Cryostorage, liquid nitrogen, refrigeration, and ultra-low temperature freezer applications