missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FITC/5-FAM/6-FAM Recombinant Rabbit Monoclonal Antibody (4L8R5)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA555271

Product Code. 30132058

  • € 436.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen Sequence: FITC conjugated to KLH.

FITC (Fluorescein Isothiocyanate), 5-FAM (5-Carboxyfluorescein), and 6-FAM (6-Carboxyfluorescein) are commonly used gene targets for fluorescent labeling in molecular biology research. These dyes belong to the fluorescein family and emit a green fluorescence when excited by specific wavelengths of light. FITC, 5-FAM, and 6-FAM are utilized in various techniques such as fluorescence in situ hybridization (FISH), flow cytometry, immunofluorescence, and DNA sequencing. By attaching these dyes to gene-specific probes or primers, researchers can visualize and detect gene expression levels, identify specific DNA or RNA sequences, and study cellular processes. The compatibility of FITC, 5-FAM, and 6-FAM with different imaging systems and their bright green fluorescence make them valuable tools in molecular biology, aiding in the understanding of gene function and cellular dynamics.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FITC/5-FAM/6-FAM
Recombinant Monoclonal
1 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Synthetic Peptide.
100 μL
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
4L8R5
Unconjugated
Rabbit
Affinity chromatography
RUO
Chemical
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.