missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FITC Antibody, Alexa Fluor™ 350 , Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB600-1273AF350
This item is not returnable.
View return policy
Description
FITC Polyclonal antibody specifically detects FITC in Rat, Nonspecies specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| FITC | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Rat, Non-species specific | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 350 | |
| Fluorescein Isothiocyanate | |
| FITC conjugated to KLH. | |
| 1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction