missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FITC Antibody, Janelia Fluor™ 646 , Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NB100-64617JF646
This item is not returnable.
View return policy
Description
FITC Polyclonal antibody specifically detects FITC in Nonspecies specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, ELISA
Specifications
| FITC | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Sheep | |
| Immunogen affinity purified | |
| RUO | |
| Non-species specific | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| Janelia Fluor 646 | |
| Fluorescein Isothiocyanate | |
| Pooled antisera from sheep hyperimmunized with fluorescein isothiocyanate (Isomer I) | |
| 1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction