missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FIV gp95 Antibody (SU1-30), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64378H
This item is not returnable.
View return policy
Description
FIV gp95 Monoclonal antibody specifically detects FIV gp95 in Virus samples. It is validated for Western Blot
Specifications
| FIV gp95 | |
| Monoclonal | |
| HRP | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot | |
| SU1-30 | |
| PBS | |
| Made to FIV gp95 | |
| 0.125 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction