missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FIV gp95 Antibody (SU1-30), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64378JF549
This item is not returnable.
View return policy
Description
FIV gp95 Monoclonal antibody specifically detects FIV gp95 in Virus samples. It is validated for Western Blot, Flow Cytometry
Specifications
| FIV gp95 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot, Flow Cytometry | |
| SU1-30 | |
| 50mM Sodium Borate | |
| Made to FIV gp95 | |
| 0.125 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion