missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP10 Antibody (2D4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00060681-M13
This item is not returnable.
View return policy
Description
FKBP10 Monoclonal antibody specifically detects FKBP10 in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
Specifications
| FKBP10 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| 65 kDa FK506-binding protein, 65 kDa FKBP, EC 5.2.1.8, FK506 binding protein 10 (65 kDa), FK506 binding protein 10, 65 kDa, FK506-binding protein 10, FKBP-10, FKBP6, FKBP-65, FKBP65PPIase FKBP10, FLJ20683, FLJ22041, FLJ23833, hFKBP65, Immunophilin FKBP65, OI6, peptidyl-prolyl cis-trans isomerase FKBP10, PPIASE, Rotamase | |
| FKBP10 (NP_068758, 377 a.a. ∽ 470 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. ADVVEIRTLSRPSETCNETTKLGDFVRYHYNCSLLDGTQLFTSHDYGAPQEATLGANKVIEGLDTGLQGMCVGERRQLIVPPHLAHGESGARGV | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Sandwich ELISA | |
| 2D4 | |
| Western Blot, ELISA, Sandwich ELISA | |
| NP_068758 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 60681 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction