missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP12 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97584G
This item is not returnable.
View return policy
Description
FKBP12 Polyclonal antibody specifically detects FKBP12 in Human samples. It is validated for Western Blot
Specifications
| FKBP12 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 488 | |
| 12 kDa FKBP, EC 5.2.1.8, FK506 binding protein 1A, 12kDa, FK506-binding protein 1A, FKBP1, FKBP12C, FKBP12FKBP12-Exip3, FKBP-12T-cell, 12-kD, FKBP-1A, peptidyl-prolyl cis-trans isomerase FKBP1A, PPIase FKBP1A, protein kinase C inhibitor 2, rotamase | |
| Produced in rabbits immunized with E. coli-derived Human FKBP12 fragment. | |
| 0.1 mL | |
| Autophagy, Cancer, mTOR Pathway | |
| 2280 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction