missing translation for 'onlineSavingsMsg'
Learn More

FKBP12 Antibody (OTI3B3), CoraFluor™ 1, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-70741CL1

Product Code. 30145892

  • € 1220.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

FKBP12 Monoclonal antibody specifically detects FKBP12 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FKBP12
Monoclonal
CoraFluor 1
12 kDa FKBP, EC 5.2.1.8, FK506 binding protein 1A, 12kDa, FK506-binding protein 1A, FKBP1, FKBP12C, FKBP12FKBP12-Exip3, FKBP-12T-cell, 12-kD, FKBP-1A, peptidyl-prolyl cis-trans isomerase FKBP1A, PPIase FKBP1A, protein kinase C inhibitor 2, rotamase
Full length human recombinant protein of human FKBP1A (NP_000792) produced in HEK293T cell.
0.1 mL
Autophagy, Cancer, mTOR Pathway
2280
Store at 4°C in the dark. Do not freeze.
IgG2b
Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF
OTI3B3
PBS
Mouse
Immunogen affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.