missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP12 Antibody (OTI3B3), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70741PEATT594
This item is not returnable.
View return policy
Description
FKBP12 Monoclonal specifically detects FKBP12 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| FKBP12 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| FKBP1A | |
| Full length recombinant protein of human FKBP12 (NP_000792) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Flow Cytometry | |
| OTI3B3 | |
| Flow Cytometry | |
| 12 kDa FKBP, EC 5.2.1.8, FK506 binding protein 1A, 12kDa, FK506-binding protein 1A, FKBP1, FKBP12C, FKBP12FKBP12-Exip3, FKBP-12T-cell, 12-kD, FKBP-1A, peptidyl-prolyl cis-trans isomerase FKBP1A, PPIase FKBP1A, protein kinase C inhibitor 2, rotamase | |
| Mouse | |
| Affinity Purified | |
| Autophagy, Cancer, mTOR Pathway | |
| 2280.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction