missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FKBP12 Polyclonal Antibody

Product Code. 11511442
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11511442 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11511442 Supplier Invitrogen™ Supplier No. PA1026A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA1-026A detects FK506 binding protein 12 kDa (FKBP12) in human and mouse tissues. PA1-026A has been successfully used in Western blot, immunohistochemistry, and immunoprecipitation procedures. By Western blot, this antibody detects a 12 kDa protein representing FKBP12 from rat heart homogenate. The PA1-026A immunogen is a synthetic peptide corresponding to the N-terminal residues G(1) V Q V E T I S P G D G R(13) of human FKBP12. PA1-026A immunizing peptide (Cat. # PEP-012) is available for use in neutralization and control experiments.

FKBP12 is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. The protein is a cis-trans prolyl isomerase that binds the immunosuppressants FK506 and rapamycin. It interacts with several intracellular signal transduction proteins including type I TGF-beta receptor. It also interacts with multiple intracellular calcium release channels, and coordinates multi-protein complex formation of the tetrameric skeletal muscle ryanodine receptor. In mouse, deletion of this homologous gene causes congenital heart disorder known as noncompaction of left ventricular myocardium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FKBP12
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene FKBP1A
Gene Accession No. P26883, P62942
Gene Alias 12 kDa FK506-binding protein; 12 kDa FKBP; calstabin 1; calstabin-1; EC 5.2.1.8; FK506 binding protein 1a; FK506 binding protein 1A, 12kDa; FK506 binding protein 2 (13 kDa); FK506 binding protein12; FK506-binding protein 1; FK506-binding protein 1 (12kD); FK506-binding protein 12; FK506-binding protein 1a; FK506-binding protein 1A (12kD); FK506-binding protein, T-cell, 12-kD; Fkbp; FKBP1; FKBP12; FKBP-12; FKBP12-Exip3; Fkbp1a; FKBP-1A; Fkbp2; Immunophilin FKBP12; peptidyl-prolyl cis-trans isomerase FKBP1A; PKC12; PKCI2; PPIASE; PPIase FKBP1A; protein kinase C inhibitor 2; rotamase
Gene Symbols FKBP1A
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the N-terminal residues G(1) V Q V E T I S P G D G R(13) of human FKBP12.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14225, 2280
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.