missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP15 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77352AF700
This item is not returnable.
View return policy
Description
FKBP15 Polyclonal antibody specifically detects FKBP15 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| FKBP15 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| FK506 binding protein 15, 133kDa, FK506-binding protein 133kDa, FK506-binding protein 15, FKBP133, FKBP-133, FKBP-15,133 kDa FKBP, PPP1R76,133 kDa FK506-binding protein, regulatory subunit 76, WAFL | |
| Antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus of human FKBP15. The immunogen is located within amino acids 30 - 80 of FKBP15. Amino Acid Squence: YTAPKQPKKGQGTAATG | |
| 0.1 mL | |
| Signal Transduction | |
| 23307 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction