missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11700-1-AP-150UL
This item is not returnable.
View return policy
Description
FKBP2 is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. This protein is a cis-trans prolyl isomerase that binds the immunosuppressants FK506 and rapamycin. It is thought to function as an ER chaperone and may also act as a component of membrane cytoskeletal scaffolds.Specifications
| FKBP2 | |
| Polyclonal | |
| Unconjugated | |
| FKBP2 | |
| 13 kDa FK506 binding protein, 13 kDa FKBP, FK506 binding protein 2, FK506 binding protein 2, 13kDa, FKBP 13, FKBP 2, FKBP13, FKBP2, Immunophilin FKBP13, PPIase, PPIase FKBP2, Rotamase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14227, 2286, 293702 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P26885, P45878 | |
| FKBP2 | |
| FKBP2 Fusion Protein Ag2255 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction