missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11700-1-AP-20UL
This item is not returnable.
View return policy
Description
FKBP2 is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. This protein is a cis-trans prolyl isomerase that binds the immunosuppressants FK506 and rapamycin. It is thought to function as an ER chaperone and may also act as a component of membrane cytoskeletal scaffolds.Specifications
| FKBP2 | |
| Polyclonal | |
| Unconjugated | |
| FKBP2 | |
| 13 kDa FK506 binding protein, 13 kDa FKBP, FK506 binding protein 2, FK506 binding protein 2, 13kDa, FKBP 13, FKBP 2, FKBP13, FKBP2, Immunophilin FKBP13, PPIase, PPIase FKBP2, Rotamase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14227, 2286, 293702 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P26885, P45878 | |
| FKBP2 | |
| FKBP2 Fusion Protein Ag2255 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur