missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP38 Antibody (SR1916), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22302-100ul
This item is not returnable.
View return policy
Description
FKBP38 Monoclonal antibody specifically detects FKBP38 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence
Specifications
| FKBP38 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| SR1916 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| 38 kDa FK506-binding protein, EC 5.2.1.8,38 kDa FKBP, FK506 binding protein 8, 38kDa, FK506-binding protein 8, FK506-binding protein 8 (38kD), FKBP-38, FKBP38PPIase FKBP8, FKBP-8, FKBPr38, hFKBP38, peptidyl-prolyl cis-trans isomerase FKBP8, rotamase | |
| A synthesized peptide derived from human FKBP38 (Uniprot #: Q14318) | |
| 100 μg | |
| Angiogenesis, Apoptosis, Cancer, Cellular Markers, Core ESC Like Genes, Glia Markers, Growth and Development, Mitotic Regulators, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Tumor Suppressors | |
| 23770 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction