missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP52/FKBP4 Antibody (Hi52C), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-96874G
This item is not returnable.
View return policy
Description
FKBP52/FKBP4 Monoclonal specifically detects FKBP52/FKBP4 in Human, Mouse, Rat, Canine, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| FKBP52/FKBP4 | |
| Monoclonal | |
| DyLight 488 | |
| Q02790 | |
| FKBP4 | |
| Synthetic peptide corresponding to the residues of human FKBP52 | |
| 0.1 mL | |
| Membrane Trafficking and Chaperones, Stem Cell Markers | |
| 2288 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Hi52C | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| 51 kDa FK506-binding protein, 52 kDa FKBP, 59 kDa immunophilin, EC 5.2.1.8, FK506 binding protein 4, 59kDa, FK506-binding protein 4, FK506-binding protein 4 (59kD), FKBP-4, FKBP51, FKBP-52, FKBP52T-cell FK506-binding protein, 59kD, FKBP5952 kDa FK506-binding protein, HBI, HSP binding immunophilin, Hsp56, HSP-binding immunophilin, Immunophilin FKBP52, p52, p59, peptidylprolyl cis-trans isomerase, peptidyl-prolyl cis-trans isomerase FKBP4, PPIase, PPIase FKBP4, Rotamase | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Detects approx 52kDa. Heavy chain migrates close to FKBP52 on SDS PAGE. | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction