missing translation for 'onlineSavingsMsg'
Learn More

FKBP52/FKBP4 Antibody (KN382/EC1), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 3 publications

€ 161.00 - € 771.00

Specifications

Antigen FKBP52/FKBP4
Clone KN382/EC1
Dilution Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 10 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500
Applications Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18028139
View Documents
Novus Biologicals
NBP1-97465-0.025MG
0.025 mg
€ 161.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18192740
View Documents
Novus Biologicals
NBP1-97465-0.05MG
0.05 mg
€ 391.00
0.05mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18026877
View Documents
Novus Biologicals
NBP1-97465-0.2MG
0.2 mg
€ 771.00
0.20mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FKBP52/FKBP4 Monoclonal specifically detects FKBP52/FKBP4 in Human, Mouse, Bovine, Guinea Pig, Hamster, Mustelid, Primate, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

FKBP52/FKBP4
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 10 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500
Monoclonal
Purified
RUO
Guinea Pig, Primate, Human, Mouse, Hamster, Bovine, Rabbit, Mustelid
51 kDa FK506-binding protein, 52 kDa FKBP, 59 kDa immunophilin, EC 5.2.1.8, FK506 binding protein 4, 59kDa, FK506-binding protein 4, FK506-binding protein 4 (59kD), FKBP-4, FKBP51, FKBP-52, FKBP52T-cell FK506-binding protein, 59kD, FKBP5952 kDa FK506-binding protein, HBI, HSP binding immunophilin, Hsp56, HSP-binding immunophilin, Immunophilin FKBP52, p52, p59, peptidylprolyl cis-trans isomerase, peptidyl-prolyl cis-trans isomerase FKBP4, PPIase, PPIase FKBP4, Rotamase
FKBP4
IgG1
Protein G purified
KN382/EC1
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunohistochemistry
Unconjugated
Mouse
Membrane Trafficking and Chaperones, Stem Cell Markers
PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
2288
Partially purified 9S rabbit uterine progesterone receptor.
Primary
Store at -20C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.