missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP6 Antibody (9B7), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42645F
This item is not returnable.
View return policy
Description
FKBP6 Monoclonal specifically detects FKBP6 in Human samples. It is validated for ELISA.
Specifications
| FKBP6 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| FKBP6 | |
| Recombinant human FKBP6 (1-327aa) purified from E. coli | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 9B7 | |
| ELISA | |
| 36 kDa FKBP, EC 5.2.1.8, FK506 binding protein 6, 36kDa, FK506-binding protein 6, FK506-binding protein 6 (36kD), FKBP-36, FKBP36MGC87179, FKBP-6, Immunophilin FKBP36, peptidyl-prolyl cis-trans isomerase FKBP6,36 kDa FK506-binding protein, PPIase, PPIase FKBP6, rotamase | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 8468 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction