missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP7 Antibody (005), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90201CL1
This item is not returnable.
View return policy
Description
FKBP7 Monoclonal antibody specifically detects FKBP7 in Human samples. It is validated for ELISA
Specifications
| FKBP7 | |
| Monoclonal | |
| CoraFluor 1 | |
| EC 5.2.1.8, FK506 binding protein 7, FK506-binding protein 7FK506-binding protein 23,23 kDa FK506-binding protein, FKBP-23, FKBP23PPIase, FKBP-7, MGC9420, peptidyl-prolyl cis-trans isomerase FKBP7,23 kDa FKBP, PPIase FKBP7, rotamase | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human FKBP7 (Uniprot#: Q9Y680-2; Met1-Gln218). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 005 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 51661 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction