missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FKBP7 Polyclonal Antibody

Rabbit Polyclonal Antibody
€ 184.00 - € 485.00
Specifications
| Antigen | FKBP7 |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17720534
![]() |
Invitrogen™
PA5119120 |
200 μL |
€ 485.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17713384
|
Invitrogen™
PA5121983 |
20 μL |
€ 184.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FKBP7 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
The protein encoded by this gene belongs to the FKBP-type peptidyl-prolyl cis/trans isomerase (PPIase) family. Members of this family exhibit PPIase activity and function as molecular chaperones. A similar protein in mouse is located in the endoplasmic reticulum and binds calcium.Specifications
| FKBP7 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| FKBP7 | |
| 23 kDa FK506-binding protein; 23 kDa FKBP; 23kDa; FK506 binding protein 7; FK506-binding protein 23; FK506-binding protein 7; FKBP23; FKBP-23; Fkbp7; FKBP-7; Peptidyl-prolyl cis-trans isomerase FKBP7; PPIase; PPIase FKBP7; rotamase; UNQ670/PRO1304 | |
| FKBP7 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9Y680 | |
| 51661 | |
| Recombinant protein FK506 Binding Protein 7. The antigen corresponds to amino acid range 24-217 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title