missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FKBP9 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA552952

Product Code. 15750901

  • € 582.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: TGMDQALVGM CVNERRFVKI PPKLAYGNEG VSGVIPPNSV LHFDVLLMDI WNSEDQVQIH TYFKPPSCPR TIQVSDF Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 92%.

FKBP9 is a PPIase which accelerate the folding of proteins during protein synthesis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FKBP9
Polyclonal
Unconjugated
FKBP9
1810014L23; 63 kDa FK506-binding protein; 63 kDa FKBP; AW549641; FK506 binding protein 9; FK506 binding protein 9, 63 kDa; FK506-binding protein 9; FKBP60; FKBP63; FKBP-63; FKBP65RS; Fkbp9; FKBP-9; Peptidyl-prolyl cis-trans isomerase FKBP9; PPIase; PPIase FKBP9; rotamase
Rabbit
Antigen affinity chromatography
RUO
11328
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.10 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O95302
FKBP9
Recombinant protein corresponding to Human FKBP9. Recombinant protein control fragment (Product #RP-90742).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.