Learn More
FKBPL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539168
This item is not returnable.
View return policy
Description
The protein encoded by this gene has similarity to the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. The encoded protein is thought to have a potential role in the induced radioresistance. Also it appears to have some involvement in the control of the cell cycle. [provided by RefSeq, Jul 2008].
Specifications
| FKBPL | |
| Polyclonal | |
| Unconjugated | |
| FKBPL | |
| DIR1, NG7, WISP39, FK506-binding protein like, FK506-binding protein-like, WAF-1/CIP1 stabilizing protein 39 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 406168, 56299, 63943 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UIM3, O35450, Q6MG81 | |
| FKBPL | |
| A synthetic peptide derived from the internal region of human FKBPL | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |