Learn More
Liebherr™ FKvsl Series Premium Forced Air Refrigerators
Product Code. 15766828 Shop All Liebherr Products
Click to view available options
Capacity (Metric):
240 L
250 L
572 L
Lid:
Double-glazed glass door
Solid door
Unit Size:
Each
Product Code. 15766828
Brand: Liebherr™ FKvsl 2610
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Maintain a uniform temperature and consistent humidity level throughout the interior. Liebherr™ FKvsl Series Premium Forced Air Refrigerators are made of premium stainless steel, ergonomically designed and easy to clean. Available in various sizes with either a solid steel or glazed glass door.
  • Dynamic forced air cooling: Cools fresh items quickly and features a door-operated fan switch that prevents warm air from the outside from entering the appliance when open
  • Stainless steel housing: Constructed of a highly robust alloy steel that is extremely hygienic and ideal for the professional sector
  • Efficient refrigeration system: Reduces energy consumption and operating costs with premium-quality compressors, condensers, fans and other components
  • Digital temperature display: Lets users easily view the interior temperature to the exact degree from the outside
  • SwingLine door with bar handle and reversible hinges: Stylishly contoured, ergonomically designed for any space, easy to clean and easy to open even with constant use
  • Seamless plastic inner container: Thermoformed one-piece interior is extremely easy to clean; robust and long-lasting; and made of environmentally friendly, recyclable polystyrene
  • Adjustable grid shelves: Robust shelves are height-adjustable so the interior layout can be varied as required for items of all sizes stored upright or laying down
  • Environmentally friendly refrigerant: Minimal greenhouse effect and no ozone depletion potential with outstanding energy efficiency
Specifications
Type General Purpose Refrigerator
Capacity (Metric) 240 L
Temperature Range 1°C to 15°C
Refrigerant R 600a
Door Style Self-closing SwingLine
No. of Doors 1
Hinge Side Right Interchangeable
Shelves 5 (4 Adjustable) Grid
Capacity (Metric) Shelf 45 kg
Exterior Finish Stainless Steel
Interior Thermoformed One-piece White Plastic
Display Digital, External
Color Silver
Control Type Mechanical
Includes Adjustable feet
Defrost Automatic
Lid Solid door
Lighting Ceiling
Model FKvsl 2610
Access Security Key Lock
Dimensions (L x W x H) Exterior 61 x 60 x 125 cm
Depth (Metric) Exterior 61 cm
Height (Metric) Exterior 125 cm
Width (Metric) Exterior 60 cm
Amperage 1.0 A
Voltage 220/240 V
Electrical Requirements 220 to 240 V 50 Hz 1.0 A
Frequency 50 Hz
Energy Usage 0.786 kWh / 24 hr.
Shipping Weight (Metric) 51 kg
Weight (Metric) 46 kg