missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flagellin Antibody (06), FITC , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05967F
This item is not returnable.
View return policy
Description
Flagellin Monoclonal antibody specifically detects Flagellin in Bacteria samples. It is validated for ELISA
Specifications
| Flagellin | |
| Monoclonal | |
| FITC | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Bacteria | |
| Purified |
| ELISA | |
| 06 | |
| PBS | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Listeria monocytogenes Listeria monocytogenes Flagellin (YP_013330.1; Met1-Ser287). | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction