missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ FLAP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30917-25ug
This item is not returnable.
View return policy
Description
FLAP Polyclonal antibody specifically detects FLAP in Human samples. It is validated for Western Blot
Specifications
| FLAP | |
| Polyclonal | |
| Western Blot : 1:400 | |
| arachidonate 5-lipoxygenase-activating protein, FLAPMK-886-binding protein | |
| Synthetic peptide derived from human FLAP protein. | |
| 25 μg | |
| Immunology, Inflammation | |
| 241 | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction